missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucyl-Diphenylborinat, 95 %, Thermo Scientific™

Catalog Number p-7036788
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
124383-92-2
C18H22BNO2
295.189
MFCD22125121
VNPIANARMZOPIS-YOEHRIQHSA-N
diphenylboranyl 2s,3s-2-amino-3-methylpentanoate, l-isoleucyl diphenylborinate
99738123
Diphenylboranyl (2S,3S)-2-amino-3-methylpentanoat
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(C(C)CC)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
124383-92-2
C18H22BNO2
295.189
MFCD22125121
VNPIANARMZOPIS-YOEHRIQHSA-N
diphenylboranyl 2s,3s-2-amino-3-methylpentanoate, l-isoleucyl diphenylborinate
99738123
Diphenylboranyl (2S,3S)-2-amino-3-methylpentanoat
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(C(C)CC)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material L-Isoleucyl diphenylborinate
Menge 250 mg
Formelmasse 295.19
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'