missing translation for 'onlineSavingsMsg'
Learn More

L-Leucyldiphenylborinat, 95 %, Thermo Scientific™

Catalog Number p-7037197
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
116628-53-6
C18H22BNO2
295.189
MFCD22125182
HEFYWEFHUCZFOZ-KRWDZBQOSA-N
diphenylboranyl 2s-2-amino-4-methylpentanoate, l-leucyl diphenylborinate
99738149
Diphenylboranyl (2S)-2-amino-4-methylpentanoat
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CC(C)C)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
116628-53-6
C18H22BNO2
295.189
MFCD22125182
HEFYWEFHUCZFOZ-KRWDZBQOSA-N
diphenylboranyl 2s-2-amino-4-methylpentanoate, l-leucyl diphenylborinate
99738149
Diphenylboranyl (2S)-2-amino-4-methylpentanoat
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CC(C)C)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material L-Leucyl diphenylborinate
Menge 250 mg
Formelmasse 295.19
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'