missing translation for 'onlineSavingsMsg'
Learn More

L-Menthol, 99 %, Thermo Scientific Chemicals

Catalog Number p-7080785
Click to view available options
Menge:
50 g
250 g
1000 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2216-51-5
C10H20O
156.269
MFCD00062979
NOOLISFMXDJSKH-KXUCPTDWSA-N
l-menthol, --menthol, levomenthol, menthomenthol, l---menthol, hexahydrothymol, menthacamphor, peppermint camphor, 1r,2s,5r-2-isopropyl-5-methylcyclohexanol, u.s.p. menthol
16666
CHEBI:15409
(1R,2S,5R)-5-Methyl-2-propan-2-ylcyclohexan-1-ol
CC1CCC(C(C1)O)C(C)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2216-51-5
C10H20O
156.269
MFCD00062979
NOOLISFMXDJSKH-KXUCPTDWSA-N
l-menthol, --menthol, levomenthol, menthomenthol, l---menthol, hexahydrothymol, menthacamphor, peppermint camphor, 1r,2s,5r-2-isopropyl-5-methylcyclohexanol, u.s.p. menthol
16666
CHEBI:15409
(1R,2S,5R)-5-Methyl-2-propan-2-ylcyclohexan-1-ol
CC1CCC(C(C1)O)C(C)C

L-Menthol is used as a cooling agent that strongly activates TRPM8.(Transient Receptor Potential Cation Channel, Subfamily M, Member 8 is a Protein Coding gene). It is used as analgesic (topical), antipruritic agent. It is used as: refreshing agent, food flavor, cool and antipruritic drug, carminative drug. Menthol crystals is used for pers onal care and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Löslichkeit
Löslich in 100 % Ethanol, Methanol (100 mg/ml), Äther und Chloroform. Unlöslich in Wasser.

Hinweise
Bei Raumtemperatur lagern. Nicht kompatibel mit starken Oxidationsmitteln, Phenolen, Kaliumpermanganat, Chromtrioxid, Chlorhydrat, Thymol, Pyrogallol, Resorcin, Campher, Butylchlorhydrat, Betanaphthol, Exalgin.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material L-Menthol
Schmelzpunkt 42°C to 45°C
Dichte 0.89
Siedepunkt 212°C to 216°C
Flammpunkt 93°C (199°F)
Menge 50 g
Beilstein 1902293
Merck Index 14,5837
Löslichkeitsinformationen Soluble in 100% ethanol,methanol (100mg/ml),ether,and chloroform. Insoluble in water.
Optische Drehung −50° (c=10 in ethanol)
Formelmasse 156.27
Reinheit (%) 99%
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'