missing translation for 'onlineSavingsMsg'
Learn More

L-Nipekotische Säure, 96+%, Thermo Scientific Chemicals

Catalog Number p-7038552
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
59045-82-8
C6H11NO2
129.159
MFCD01630807
XJLSEXAGTJCILF-YFKPBYRVSA-N
s-piperidine-3-carboxylic acid, s-+-nipecotic acid, s-nipecotic acid, 3s-piperidine-3-carboxylic acid, +-nipecotic acid, s-+-3-piperidinecarboxylic acid, 3s-nipecotic acid, l +-nipecotic acid, s-hexahydronicotinic acid, +-hexahydronicotinic acid
6575389
CHEBI:222169
(3S)-Piperidin-3-carbonsäure
C1CC(CNC1)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
59045-82-8
C6H11NO2
129.159
MFCD01630807
XJLSEXAGTJCILF-YFKPBYRVSA-N
s-piperidine-3-carboxylic acid, s-+-nipecotic acid, s-nipecotic acid, 3s-piperidine-3-carboxylic acid, +-nipecotic acid, s-+-3-piperidinecarboxylic acid, 3s-nipecotic acid, l +-nipecotic acid, s-hexahydronicotinic acid, +-hexahydronicotinic acid
6575389
CHEBI:222169
(3S)-Piperidin-3-carbonsäure
C1CC(CNC1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material L-Nipecotic acid
Schmelzpunkt 253°C to 257°C
Formelmasse 129.16
Reinheit (%) ≥96%
Menge 1 g
Optische Drehung +5° (c=1 in Water)

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'