Learn More
L-Ornithine Hydrochloride-13C5, TRC
Product Code. 17987524
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17987524 5 mg 5mg
17939074 1 mg 1mg
17948664 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17987524 Supplier Toronto Research Chemicals Supplier No. TRCO6955515MG
 Restricted Item
Verfügbar ab: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula 13C5H12N2O2 • HCl
Molecular Weight (g/mol) 137.123646
Synonym (S)-2,5-Diaminopentanoic Acid Hydrochloride-13C5, L-Ornithine Monohydrochloride-13C5, L-(+)-Ornithine Monohydrochloride-13C5, (S)-α,δ-Diaminovaleric Acid-13C5, 5-Amino-L-norvaline-13C5;
SMILES N[13C@@]([13C]([H])([H])[13C]([H])([H])[13C]([H])([H])N)([H])[13C](O)=O.Cl
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m1./s1/i1+1,2+1,3+1,4+1,5+1;
Percent Purity >95%
Chemical Name or Material L-Ornithine Hydrochloride-13C5
Grade HPLC