missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanyl-Diphenylborinat, 95 %, Thermo Scientific™

Catalog Number p-7036824
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
88879-35-0
C21H20BNO2
329.206
MFCD22380721
KHDSFZVARCBQMJ-FQEVSTJZSA-N
l-phenylalanyl diphenylborinate, diphenylboranyl 2s-2-amino-3-phenylpropanoate, l-phenylalanine diphenylboryl ester, diphenylborinic acid l-phenylalanyl ester
73996109
Diphenylboranyl-(2S)-2-amino-3-phenylpropanoat
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CC3=CC=CC=C3)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
88879-35-0
C21H20BNO2
329.206
MFCD22380721
KHDSFZVARCBQMJ-FQEVSTJZSA-N
l-phenylalanyl diphenylborinate, diphenylboranyl 2s-2-amino-3-phenylpropanoate, l-phenylalanine diphenylboryl ester, diphenylborinic acid l-phenylalanyl ester
73996109
Diphenylboranyl-(2S)-2-amino-3-phenylpropanoat
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CC3=CC=CC=C3)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material L-Phenylalanyl diphenylborinate
Menge 250 mg
Formelmasse 329.21
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'