Learn More
L1CAM Mouse anti-Human, Biotin, Clone: 3H7B9, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech BIOTIN-67115-100UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The L1CAM antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human L1CAM. This antibody recognizes human, pig, mouse, rat antigen. The L1CAM antibody has been validated for the following applications: IHC analysis. This antibody is biotin.
Spezifikation
L1CAM | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Rat, Mouse, Human | |
IgG1 |
Immunohistochemistry | |
3H7B9 | |
Biotin | |
100 μL | |
3897 | |
Liquid |