Learn More
Epredia™ Lab Vision™ Lambda Light Chain Ab-4 (Cocktail) Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Lambda Light Chain Ab-4 (Cocktail), Mouse Monoclonal Antibody provides accurate, reproducible results.
391.00 €
Spezifikation
Antigen | Lambda Light Chain Ab-4 (Cocktail) |
---|---|
Klon | LAM03 + HP6054 |
Anwendungen | Immunohistochemistry (Paraffin) |
Klassifikation | Monoclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Form | Menge | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Form | Menge | Preis | Menge & Verfügbarkeit | ||||
12669136
|
Epredia™
MS-1158-P |
Concentrated | 1 mL |
391.00 €
1 Milliliter |
Verfügbar ab: 12-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||
Beschreibung
Antibody to the λ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Mouse
Clone: LAM03 + HP6054
Isotype: IgG1/κ + IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Purified human Lambda light chain, and purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads
Molecular Weight: 25kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic Ab-4 cocktail is especially designed for sensitive detection of lambda light chain of immunoglobulin.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Spezifikation
Lambda Light Chain Ab-4 (Cocktail) | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
IgG2a κ |
LAM03 + HP6054 | |
Monoclonal | |
Mouse | |
Hematopoietic | |
Purified human lambda light chain, and purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads | |
Primary |