missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin alpha-2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA549929
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total Laminin alpha-2 protein.
Laminin alpha-2 Polyclonal antibody specifically detects Laminin alpha-2 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
Laminin alpha-2 | |
Polyclonal | |
Unconjugated | |
Lama2 | |
5830440B04; dy; dystrophia muscularis; Lama2; laminin alpha-2; laminin M; Laminin M chain; laminin subunit alpha 2; laminin subunit alpha-2; laminin, alpha 2; laminin-12 subunit alpha; Laminin-2 subunit alpha; laminin-4 subunit alpha; LAMM; mer; merosin; Merosin heavy chain; mKIAA4087 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16773, 3908 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P24043, Q60675 | |
Lama2 | |
Synthesized peptide derived from internal of human LAMA2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |