missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody Kit
LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody Kit
Marke: Thermo Scientific™ PV5130
Weitere Details : Netto-Gewicht : 0.09000kg
Beschreibung
The p53 tumor suppressor protein is an essential component of many different signaling pathways. The LanthaScreen™ antibody detects the phosphorylation of p53 at Ser15 by a range of kinases including DNA-PK, ATM, and ATR.For use in combination with the Fluorescein-p53 (Ser15) peptide substrate (PV5132) to evaluate inhibitors in medium to high throughput screening applications.
Spezifikation
Serine/threonine kinase | |
0.53mg/mL | |
150kDa | |
RUO, RUO | |
Primary | |
Terbium-labeled anti-phospho-p53 [pSer15] antibody in HEPES buffered saline. Store at -20°C. | |
Biochemical terbium phospho-specific activity | |
340⁄490, 520 | |
Dry Ice | |
Hu | |
Serine⁄Threonine Kinases | |
LanthaScreen™ |
Kinase Assay, Kinase Assay | |
Tb (Terbium)/FITC/Lanthanide Chelates | |
25 μg | |
Oncology and Genetic Disease Research | |
1 Kit(s) | |
150000 Da | |
384-well Plate | |
Human | |
End Point | |
DNAPK | |
TR-FRET |
For Research Use Only. Not for use in diagnostic procedures.