missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAPTM4A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556731
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EVTHPNSMPA VNIQYEVIGN YYSSERMADN Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
LAPTM4A Polyclonal antibody specifically detects LAPTM4A in Human samples. It is validated for Immunohistochemistry.Spezifikation
LAPTM4A | |
Polyclonal | |
Unconjugated | |
LAPTM4A | |
AA286466; golgi 4-transmembrane spanning transporter; golgi 4-transmembrane-spanning transporter; golgi 4-transmembrane-spanning transporter MTP; HUMORF13; KIAA0108; LAPTM4; LAPTM4A; lysosomal protein transmembrane 4 alpha; lysosomal-associated protein transmembrane 4 alpha; lysosomal-associated protein transmembrane 4A; lysosomal-associated transmembrane protein 4A; lysosomal-associated transmembrane protein 4-like protein; MBNT; membrane nucleoside transporter; membrane transporter protein; mKIAA0108; MTP; Mtrp; similar to lysosomal-associated protein transmembrane 4 alpha; transmembrane glycoprotein; transporter protein; UNQ1846/PRO3574; wu:fb57c07; wu:fj99e08 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9741 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15012 | |
LAPTM4A | |
Recombinant protein corresponding to Human LAPTM4A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only