missing translation for 'onlineSavingsMsg'
Learn More

LCTL Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 24589-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16883075

  • 112.71 € / 20 Mikroliter

Verfügbar ab
16-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes a member of family 1 glycosidases. Glycosidases are enzymes that hydrolyze glycosidic bonds and are classified into families based on primary amino acid sequence. Most members of family 1 have two conserved glutamic acid residues, which are required for enzymatic activity. The mouse ortholog of this protein has been characterized and has a domain structure of an N-terminal signal peptide, glycosidase domain, transmembrane domain, and a short cytoplasmic tail. It lacks one of the conserved glutamic acid residues important for catalysis, and its function remains to be determined. Alternative splicing results in multiple transcript variants.
Spezifikation

Spezifikation

LCTL
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6UWM7
KLG, KLPH, lactase like, Lactase like protein, LCTL
Rabbit
IgG
20 μL
-20°C
Primary
197021
Immunocytochemistry, Western Blot, Immunofluorescence
Unconjugated
LCTL
Liquid
LCTL
LCTL Fusion Protein Ag20220
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation