Learn More
LDLRAD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562622
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GAQACITLTN RTGFLCHDQR SCIPASGVCD GVRTCTHGED EDESLCRDVP QSLPHFLVAH CGDPASWI Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 68%.
LDLRAD1 belongs to the LDLR family.Spezifikation
LDLRAD1 | |
Polyclonal | |
Unconjugated | |
LDLRAD1 | |
LDLRAD1; low density lipoprotein receptor class A domain containing 1; low-density lipoprotein receptor class A domain-containing protein 1; OTTMUSG00000008594 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
388633 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T700 | |
LDLRAD1 | |
Recombinant protein corresponding to Human LDLRAD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only