missing translation for 'onlineSavingsMsg'
Learn More

LDN 57444, Tocris Bioscience™

Ubiquitin C-terminal hydrolase-L1 (UCH-L1) inhibitor

Marke:  Tocris (Bio-Techne) 399850

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413954

  • 692.44 € / 50 Milligramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Inhibitor of ubiquitin C-terminal hydrolase-L1 (UCH-L1) activity (Ki = 0.4μM). Causes cell death through the apoptosis pathway; increases levels of highly ubiquitinated proteins and decreases ubiquitin proteasome activity. Activity leads to dramatic alterations in synaptic protein distribution and spine morphology in vivo.
Spezifikation

Chemische Identifikatoren

668467-91-2
397.636
uch-l1 inhibitor, 3z-5-chloro-1-2,5-dichlorophenyl methyl-2-oxoindol-3-ylidene amino acetate
[(E)-[5-chloro-1-[(2,5-dichlorophenyl)methyl]-2-oxoindol-3-ylidene]amino] acetate
C17H11Cl3N2O3
OPQRFPHLZZPCCH-LTGZKZEYSA-N
51537051
CC(=O)ON=C1C2=C(C=CC(=C2)Cl)N(C1=O)CC3=C(C=CC(=C3)Cl)Cl

Spezifikation

LDN 57444
668467-91-2
uch-l1 inhibitor, 3z-5-chloro-1-2,5-dichlorophenyl methyl-2-oxoindol-3-ylidene amino acetate
CC(=O)ON=C1C2=C(C=CC(=C2)Cl)N(C1=O)CC3=C(C=CC(=C3)Cl)Cl
397.636
397.64
50mg
C17H11Cl3N2O3
OPQRFPHLZZPCCH-LTGZKZEYSA-N
[(E)-[5-chloro-1-[(2,5-dichlorophenyl)methyl]-2-oxoindol-3-ylidene]amino] acetate
51537051
>98%
Videos