Learn More
LEMD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553589
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EDNMKLLPVD CERKTDEFCQ AKQKAALLEL LHELYNFLAI QAGNFECGNP ENLKSKCIPV MEAQEYIANV TSSSSAKFEA ALTWILSSNK DVGIWLKGED QSELVTTVDK VVCLESAHP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.
LEMD2 Polyclonal antibody specifically detects LEMD2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
LEMD2 | |
Polyclonal | |
Unconjugated | |
LEMD2 | |
BC026588; CTRCT42; dJ482C21.1; hLEM2; LEM domain containing 2; LEM domain-containing protein 2; Lem2; LEMD2; NET25; nuclear envelope constituent; Nuclear envelope transmembrane protein 25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221496 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NC56 | |
LEMD2 | |
Recombinant protein corresponding to Human LEMD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only