missing translation for 'onlineSavingsMsg'
Learn More

Leucopterin, TRC

Product Code. 30474666
Change view
Click to view available options
Menge:
50 mg
Unit Size:
50 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30474666 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474666 Supplier Toronto Research Chemicals Supplier No. TRCL33045350MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Leucopterin
CAS 492-11-5
Summenformel C6 H5 N5 O3
Menge 50 mg
Synonym 2-Amino-5,8-dihydro-4,6,7(1H)-pteridinetrione, 2-Amino-5,8-dihydro-4,6,7(3H)-pteridinetrione, 2-Amino-3,4,5,6,7,8-hexahydropteridine-4,6,7-trione, 2-Amino-4,6,7-trihydroxypteridine, 2-Amino-4,6,7-trihydroxypyrimido[4,5-b]pyrazine, 2-Amino-5,8-dihydro-1H-pteridine-4,6,7-trione, 2-Amino-6,7-dihydroxy-3,4-dihydropteridin-4-one, 2-Aminopteridine-4,6,7-triol, Leukopterin, NSC 93740
InChI-Formel InChI=1S/C6H5N5O3/c7-6-10-2-1(3(12)11-6)8-4(13)5(14)9-2/h(H,8,13)(H4,7,9,10,11,12,14)
SMILES NC1=NC2=C(NC(=O)C(=O)N2)C(=O)N1
IUPAC-Name 2-Amino-5,8-dihydro-3H-pteridin-4,6,7-trion
Molekulargewicht (g/mol) 195.136
Formelmasse 195.039
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.