missing translation for 'onlineSavingsMsg'
Learn More

LGALS3BP Mouse anti-Human, Clone: 2D8E11, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 60066-1-IG-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16887025

  • 399.00 € / 150 Mikroliter
Verfügbar ab: 04-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP has been found elevated in the serum of patients with cancer and in those infected by the human immunodeficiency virus (HIV). It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Using fluorescence in situ hybridization the full length 90K cDNA has been localized to chromosome 17q25. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1.
Spezifikation

Spezifikation

LGALS3BP
Monoclonal
1.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q08380
LGALS3BP
LGALS3BP Fusion Protein Ag0294
150 μL
Primary
Human
Antibody
IgG2b
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
2D8E11
Unconjugated
LGALS3BP
90K, BTBD17B, Galectin 3 binding protein, LGALS3BP, M2BP, Mac 2 binding protein, MAC 2 BP, MAC2BP, Tumor associated antigen 90K
Mouse
Caprylic Acid Precipitation
RUO
3959
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote