Learn More
Huenersdorf™ Lid for 1000mL Graduated Can
Description
- Round shape
- Made of transparent polypropylene
- 122mm in diameter
Specifications
Specifications
| Color | Transparent |
| Diameter (Metric) Outer | 122mm |
| Material | Polypropylene |
| For Use With (Equipment) | 1000mL Graduated Cans |
| Quantity | 32 |
| Type | Lid |
| Style | Round |