Learn More
LRBA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553986
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SVLMVSKYRD ILEPQNERHS QSCTETGSEN ENVSLSEITP AAFSTLTTAS VEESESTSSA RRRDSGIGEE TATGLGSHVE VTPHTAPPGV SAGPDAISEV LSTLSLEVNK SPETKNDRGN DLDTKATPSV SV Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 73%.
LRBA Polyclonal antibody specifically detects LRBA in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
LRBA | |
Polyclonal | |
Unconjugated | |
LRBA | |
Beige-like protein; BGL; CDC4L; CDC4-like protein; CVID8; LAB300; LBA; Lipopolysaccharide-responsive and beige-like anchor protein; LPS responsive beige-like anchor protein; LPS-responsive vesicle trafficking, beach and anchor containing; LRBA; vesicle trafficking, beach and anchor containing | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
987 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P50851 | |
LRBA | |
Recombinant protein corresponding to Human LRBA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only