Learn More
LRCH4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558049
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LPIAGPATAP APRPLGSIQR PNSFLFRSSS QSGSGPSSPD SVLRPRRYPQ VPDEKDLMTQ LRQVLESRLQ Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.
LRCH4 is a protein that contains leucine-rich repeats (LRR) at its amino terminus and that is known to be involved in ligand binding. The carboxyl terminus may act as a membrane anchor. Identified structural elements suggest that this protein resembles a receptor.This gene encodes a protein that contains leucine-rich repeats (LRR) at its amino terminus and that is known to be involved in ligand binding. The carboxyl terminus may act as a membrane anchor. Identified structural elements suggest that the encoded protein resembles a receptor.Spezifikation
LRCH4 | |
Polyclonal | |
Unconjugated | |
LRCH4 | |
2810008P14Rik; 2900069C24Rik; AI558103; FLJ40101; FLJ46315; leucine rich repeat neuronal 4; leucine rich repeat protein 4, neuronal; leucine rich repeats and calponin homology domain containing 4; leucine-rich neuronal protein; Leucine-rich repeat and calponin homology domain-containing protein 4; Leucine-rich repeat neuronal protein 4; leucine-rich repeats and calponin homology (CH) domain containing 4; Lrch4; LRN; LRRN1; LRRN4; mFLJ00248; PP14183; SAP25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4034 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75427 | |
LRCH4 | |
Recombinant protein corresponding to Human LRCH4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only