missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
LRP2 Polyclonal specifically detects LRP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Spezifikation
Specifications
| Antigen | LRP2 |
| Anwendungen | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Zusammensetzung | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gen-Zugriffsnummer | Q96SA4 |
| Gen-Alias | calcium sensor protein, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP-2, megalin |
| Gensymbole | LRP2 |
| Wirtsspezies | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?