Learn More
LRRC10B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561558
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EIEELRELRI LALDFNKLER LPDGLCRLPR LTRLYLGGNR LLALPADFAQ LQSLRCLWIE GNFLRRF Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
LRRC10B Polyclonal antibody specifically detects LRRC10B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
LRRC10B | |
Polyclonal | |
Unconjugated | |
LRRC10B | |
Gm705; leucine rich repeat containing 10B; leucine-rich repeat-containing protein 10B; LOW QUALITY PROTEIN: leucine-rich repeat-containing protein 10B; LRRC10B; LRRC10-like protein ENSP00000367315; RGD1564983; SerdinR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
390205 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NIK2 | |
LRRC10B | |
Recombinant protein corresponding to Human LRRC10B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only