Learn More
LRRC48 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557504
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NIVDMVGLFI ENVQSLMAQC RDLENHHHEK LLEISISTLE KIVEGDLDED LPNDLRALFV DKDTIVNAVG ASHDIHLLKI DNREDELVTR INSWCTRLID RIHKDEIMRN RKRVKEINQY Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.
LRRC48 Polyclonal antibody specifically detects LRRC48 in Human samples. It is validated for Immunohistochemistry.Spezifikation
LRRC48 | |
Polyclonal | |
Unconjugated | |
DRC3 | |
CFAP134; DRC3; Dynein regulatory complex subunit 3; dynein regulatory complex subunit 3; LOW QUALITY PROTEIN: dynein regulatory complex subunit 3; leucine rich repeat containing 48; leucine-rich repeat-containing protein 48; Lrrc48; RGD1309150 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83450 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H069 | |
DRC3 | |
Recombinant protein corresponding to Human LRRC48 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only