Learn More
LRRC72 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554233
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SIAFGGKVDA SWDPKSPFKQ KPAQRVPSDF AFANNVDKTV LDDPEDAVFV RSMKRSVMTL TSMNWDTVPT REERYLEEEG TETAQMLTVT Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 64%.
LRRC72 Polyclonal antibody specifically detects LRRC72 in Human samples. It is validated for Immunohistochemistry.Spezifikation
LRRC72 | |
Polyclonal | |
Unconjugated | |
LRRC72 | |
leucine rich repeat containing 72; leucine-rich repeat-containing protein 72; leucine-rich repeat-containing protein ENSP00000371558; LRRC72 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100506049 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NJI9 | |
LRRC72 | |
Recombinant protein corresponding to Human LRRC72 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only