missing translation for 'onlineSavingsMsg'
Learn More

LRRC8C Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA556118

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 15754031

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

Immunogen sequence: LERLSFSHNK IEVLPSHLFL CNKIRYLDLS YNDIRFIPPE IGVLQSLQYF SITCNKVESL PDELYFCKKL KTLKIGKNSL SVLSPKIGNL LFLSYLDVKG NHFEILPPEL GDCRALKRAG LVVEDALFET L Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

LRRC8C is a protein coding gene. Non-essential component of the volume-regulated anion channel (VRAC, also named VSOAC channel), an anion channel required to maintain a constant cell volume in response to extracellular or intracellular osmotic changes. The VRAC channel conducts iodide better than chloride and can also conduct organic osmolytes like taurine. Plays a redundant role in the efflux of amino acids, such as aspartate and glutamate, in response to osmotic stress. Channel activity requires LRRC8A plus at least one other family member (LRRC8B, LRRC8C, LRRC8D or LRRC8E); channel characteristics depend on the precise subunit composition.
Spezifikation

Spezifikation

LRRC8C
Polyclonal
Unconjugated
LRRC8C
LRRC8C
Recombit protein corresponding to Human LRRC8C
100 μL
Primary
Human
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8TDW0
Rabbit
Antigen affinity chromatography
RUO
84230
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only