Learn More
LRRIQ3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556562
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MFHGTVTEEL TSHEEWSHYN ENIREGQKDF VFVKFNGLHL KSMENLQSCI SLRVCIFSNN FITDIHPLQS CIKLIKLDLH GN Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
The function of this protein remains unknown.Spezifikation
LRRIQ3 | |
Polyclonal | |
Unconjugated | |
LRRIQ3 | |
leucine rich repeat containing 44; leucine rich repeats and IQ motif containing 3; leucine-rich repeat and IQ domain-containing protein 3; Leucine-rich repeat-containing protein 44; leucine-rich repeats and IQ motif containing 3; LRRC44; Lrriq3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
127255, 499732 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6PVS8, Q6AYL8 | |
LRRIQ3 | |
Recombinant protein corresponding to Human LRRIQ3 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Research Use Only