missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ LTB4 Parameter Assay Kit, R&D Systems™

a 4.5 hour forward sequential competitive enzyme immunoassay designed to measure LTB4 in cell culture supernates, serum, and plasma

Marke:  R&D Systems™ SKGE006B

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412796

  • 2957.27 € / 6 Stück


Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This assay is based on the forward sequential competitive binding technique in which LTB4 present in a sample competes with a fixed amount of horseradish peroxidase (HRP)-labeled LTB4 for sites on a chicken polyclonal antibody. During the incubations, the chicken polyclonal antibody becomes bound to the rabbit anti-chicken antibody coated onto the microplate. Following a wash to remove excess conjugate and unbound sample, a substrate solution is added to the wells to determine the bound enzyme activity. Following color development, the assay is stopped, and the absorbance is read at 450 nm. The intensity of the color is inversely proportional to the concentration of LTB4 in the sample.
Spezifikation

Spezifikation

10.30 - 2500pg/mL
Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
ELISA kit
10.9pg/mL
Multi-species
6 x 96-well plates
Biotin
ELISA
1 kit
Cell Culture Supernates (50uL), Serum (50uL), EDTA Plasma (50uL), Heparin Plasma (50uL)
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Videos