missing translation for 'onlineSavingsMsg'
Learn More

Luteolin, Tocris Bioscience™

Anti-inflammatory, antioxidant and free radical scavenger

Marke:  Tocris (Bio-Techne) 287450

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13411974

  • 421.22 € / 50 Milligramm


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Anti-inflammatory, antioxidant and free radical scavenger. Inhibits LPS-induced TNF-α, IL-6 and inducible nitric oxide production and blocks NF-κB and AP-1 activation. Also inhibits TNF-α -induced COX-2 expression. Antiproliferative and chemopreventative; inhibits proliferation of Lewis lung carcinoma cells in vivo.
Spezifikation

Chemische Identifikatoren

491-70-3
286.239
luteolin, 3',4',5,7-tetrahydroxyflavone, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-4h-chromen-4-one, digitoflavone, luteolol, flacitran, luteoline, weld lake, cyanidenon 1470, salifazide
CHEBI:15864
C1=CC(=C(C=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O)O
C15H10O6
IQPNAANSBPBGFQ-UHFFFAOYSA-N
5280445
2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one

Spezifikation

Luteolin
491-70-3
luteolin, 3',4',5,7-tetrahydroxyflavone, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-4h-chromen-4-one, digitoflavone, luteolol, flacitran, luteoline, weld lake, cyanidenon 1470, salifazide
C1=CC(=C(C=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O)O
286.239
CHEBI:15864
>98%
50mg
C15H10O6
IQPNAANSBPBGFQ-UHFFFAOYSA-N
2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one
5280445
286.24
Videos