Learn More
LUZP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561241
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SERSHGHSER SHGHSERSHG HSKRSRSQGD LVDTQSDLIA TQRDLIATQK DLIATQRDLI ATQRDLIVTQ RDLVATERDL INQSGRSHGQ SERHQRYSTG K Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 25%.
LUZP4 Polyclonal antibody specifically detects LUZP4 in Human samples. It is validated for Immunohistochemistry.Spezifikation
LUZP4 | |
Polyclonal | |
Unconjugated | |
LUZP4 | |
Cancer/testis antigen 28; CT28; CT-28; CT-8; HOM-TES-85; HOM-TES-85 tumor antigen; leucine zipper protein 4; LUZP4; Tumor antigen HOM-TES-85 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51213 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P127 | |
LUZP4 | |
Recombinant protein corresponding to Human LUZP4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only