Learn More
Invitrogen™ LXR-a Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
LXR-a (Liver X Receptor alpha) ligand binding domain is a 55.9 kDa recombinant human protein (amino acids 207-447) expressed as a GST fusion protein in baculovirus-infected insect cells.
Marke: Invitrogen™ PV4657
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
LXR alpha was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Spezifikation
Nuclear Receptor Assay | |
50 μg | |
RUO - research use only | |
Liver X Receptor Alpha | |
Recombinant | |
Hormone Receptors |
10062 | |
Store in ultra-cold freezer (-68 to -85°C). | |
Recombinant, Ligand Binding Domain | |
Human | |
GST Tag |