Learn More
Ly-108 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562389
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LPLEFPAGEK VNFITWLFNE TSLAFIVPHE TKSPEIHVTN PKQGKRLNFT QSYSLQLSNL KMEDTGSYRA QISTKT Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 50%.
Ly-108 Polyclonal antibody specifically detects Ly-108 in Human samples. It is validated for Immunohistochemistry.Spezifikation
Ly-108 | |
Polyclonal | |
Unconjugated | |
SLAMF6 | |
Activating NK receptor; CD352; KAL1; KAL1b; KALI; KALIb; Ly108; Lymphocyte antigen 108; natural killer-, T- and B-cell antigen; NK-T-B-antigen; NTBA; NTB-A; NTBA receptor; RGD1561848; SF2000; SLAM family member 6; Slamf6; UNQ6123/PRO20080 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114836 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96DU3 | |
SLAMF6 | |
Recombinant protein corresponding to Human Ly-108 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only