missing translation for 'onlineSavingsMsg'
Learn More

LY 320135, Tocris Bioscience™

Selective CB1 receptor antagonist/inverse agonist

Marke:  Tocris (Bio-Techne) 238750

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13411124

  • 921.55 € / 50 Milligramm


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Potent CB 1 receptor antagonist/inverse agonist (K i = 141 nM) with greater than 70-fold selectivity over CB 2 receptors (K i > 10 μM). Structurally dissimilar from SR 141716A and AM 251. Shows weak binding to both 5-HT 2 (K i = 6.4 μM) and muscarinic receptors (K i = 2.1 μM).
Spezifikation

Chemische Identifikatoren

176977-56-3
383.403
unii-cjy03984ct, 6-methoxy-2-4-methoxyphenyl benzo b furan-3-yl 4-cyanophenyl methanone, 4-6-methoxy-2-4-methoxyphenyl benzofuran-3-carbonyl benzonitrile, 4-6-methoxy-2-4-methoxyphenyl-3-benzofuranyl carbonyl benzonitrile, 4-6-methoxy-2-4-methoxyphenyl benzofuran-3-carbonyl benzonitrile, 4-6-methoxy-2-4-methoxyphenyl-1-benzofuran-3-carbonyl benzonitrile, d0zu9z, gtpl742, benzonitrile, 4-6-methoxy-2-4-methoxyphenyl-3-benzofuranyl carbonyl, ly hplc
4-[6-methoxy-2-(4-methoxyphenyl)-1-benzofuran-3-carbonyl]benzonitrile
C24H17NO4
RYNSGDFWBJWWSZ-UHFFFAOYSA-N
5311257
COC1=CC=C(C=C1)C2=C(C3=C(O2)C=C(C=C3)OC)C(=O)C4=CC=C(C=C4)C#N

Spezifikation

LY 320135
176977-56-3
unii-cjy03984ct, 6-methoxy-2-4-methoxyphenyl benzo b furan-3-yl 4-cyanophenyl methanone, 4-6-methoxy-2-4-methoxyphenyl benzofuran-3-carbonyl benzonitrile, 4-6-methoxy-2-4-methoxyphenyl-3-benzofuranyl carbonyl benzonitrile, 4-6-methoxy-2-4-methoxyphenyl benzofuran-3-carbonyl benzonitrile, 4-6-methoxy-2-4-methoxyphenyl-1-benzofuran-3-carbonyl benzonitrile, d0zu9z, gtpl742, benzonitrile, 4-6-methoxy-2-4-methoxyphenyl-3-benzofuranyl carbonyl, ly hplc
COC1=CC=C(C=C1)C2=C(C3=C(O2)C=C(C=C3)OC)C(=O)C4=CC=C(C=C4)C#N
383.403
383.4
50mg
C24H17NO4
RYNSGDFWBJWWSZ-UHFFFAOYSA-N
4-[6-methoxy-2-(4-methoxyphenyl)-1-benzofuran-3-carbonyl]benzonitrile
5311257
>98%
Videos