missing translation for 'onlineSavingsMsg'
Learn More

LY 354740, Tocris Bioscience™

Potent and highly selective group II mGlu agonist

Marke:  Tocris (Bio-Techne) 324610

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 10mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412644

  • 265.33 € / 10 Milligramm


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Spezifikation

Chemische Identifikatoren

176199-48-7
185.179
eglumegad, eglumetad, 1s,2s,5r,6s-2-aminobicyclo 3.1.0 hexane-2,6-dicarboxylic acid, eglumegad inn, unii-onu5a67t2s, 3h eglumegad, chembl8759, onu5a67t2s, eglumegad inn, eglumegad usan
(1S,2S,5R,6S)-2-aminobicyclo[3.1.0]hexane-2,6-dicarboxylic acid
C8H11NO4
VTAARTQTOOYTES-RGDLXGNYSA-N
213056
C1CC(C2C1C2C(=O)O)(C(=O)O)N

Spezifikation

LY 354740
176199-48-7
eglumegad, eglumetad, 1s,2s,5r,6s-2-aminobicyclo 3.1.0 hexane-2,6-dicarboxylic acid, eglumegad inn, unii-onu5a67t2s, 3h eglumegad, chembl8759, onu5a67t2s, eglumegad inn, eglumegad usan
C1CC(C2C1C2C(=O)O)(C(=O)O)N
185.179
185.18
10mg
C8H11NO4
VTAARTQTOOYTES-RGDLXGNYSA-N
(1S,2S,5R,6S)-2-aminobicyclo[3.1.0]hexane-2,6-dicarboxylic acid
213056
>98%
Videos