missing translation for 'onlineSavingsMsg'
Learn More

LY 379268, Tocris Bioscience™

Highly selective group II mGlu agonist

Marke:  Tocris (Bio-Techne) 245350

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13411244

  • 1722.45 € / 50 Milligramm


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Highly selective group II mGlu receptor agonist (EC 50 values are 2.69 and 4.48 nM for hmGlu 2 and hmGlu 3 respectively) that displays > 80-fold selectivity over group I and group III receptors. Provides protection against NMDA-mediated cell death in vitro and offers almost complete protection against CA1 hippocampal damage following global ischemia in gerbils. Orally and systemically active. Caged LY 379268 is also available (Cat. No. 3332).
Spezifikation

Chemische Identifikatoren

191471-52-0
187.151
1r,4r,5s,6r-4-amino-2-oxabicyclo 3.1.0 hexane-4,6-dicarboxylic acid, d07grg, 4-amino-2-oxa-bicyclo 3.1.0 hexane-4,6-dicarboxylic acid, 1r,4r,5s,6r-4-amino-2-oxa-bicyclo 3.1.0 hexane-4,6-dicarboxylic acid, 1s,2r,5r,6r-2-amino-4-oxabicyclo 3.1.0 hexane-2,6-dicarboxylic acid, 2-oxabicyclo 3.1.0 hexane-4,6-dicarboxylic acid, 4-amino-, 1r,4r,5s,6r-rel
(1R,4R,5S,6R)-4-amino-2-oxabicyclo[3.1.0]hexane-4,6-dicarboxylic acid
C7H9NO5
YASVRZWVUGJELU-MDASVERJSA-N
10197984
C1C(C2C(C2O1)C(=O)O)(C(=O)O)N

Spezifikation

LY 379268
191471-52-0
1r,4r,5s,6r-4-amino-2-oxabicyclo 3.1.0 hexane-4,6-dicarboxylic acid, d07grg, 4-amino-2-oxa-bicyclo 3.1.0 hexane-4,6-dicarboxylic acid, 1r,4r,5s,6r-4-amino-2-oxa-bicyclo 3.1.0 hexane-4,6-dicarboxylic acid, 1s,2r,5r,6r-2-amino-4-oxabicyclo 3.1.0 hexane-2,6-dicarboxylic acid, 2-oxabicyclo 3.1.0 hexane-4,6-dicarboxylic acid, 4-amino-, 1r,4r,5s,6r-rel
C1C(C2C(C2O1)C(=O)O)(C(=O)O)N
187.151
187.15
50mg
C7H9NO5
YASVRZWVUGJELU-MDASVERJSA-N
(1R,4R,5S,6R)-4-amino-2-oxabicyclo[3.1.0]hexane-4,6-dicarboxylic acid
10197984
>99%
Videos