Learn More
LY6K Monoclonal Antibody (CL2433), Invitrogen™
Mouse Monoclonal Antibody
Marke: Thermo Scientific MA524638
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: TDEGDNRVWC HVCERENTFE CQNPRRCKWT EPYCVIAAVK IFPRFFMVAK QCSAGCAAME RPKPEEKRFL LEEPMPFFYL KCCKIRYCNL EGPPINSSVF KEYAG Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 36%. Binds to an epitope located within the peptide sequence LEEPMPFFYLKCCKI as determined by overlapping synthetic peptides.
LY6K Monoclonal antibody specifically detects LY6K in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
LY6K | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q17RY6 | |
LY6K | |
Recombinant protein corresponding to Human LY6K | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot | |
CL2433 | |
Unconjugated | |
LY6K | |
2410015A16Rik; 3110035B01Rik; cancer/testis antigen 97; CO16; CT97; FLJ35226; HSJ001348; Ly6k; ly-6K; lymphocyte antigen 6 complex, locus K; lymphocyte antigen 6 complex, locus K-like; lymphocyte antigen 6K; mLy-6K; up-regulated in lung cancer 10; URLC10 | |
Mouse | |
Protein A | |
RUO | |
54742 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only