Learn More
Corning™ Lymphocyte Serum-free Medium, KBM 551
Description
Corning’s™ Lymphocyte Serum Free Medium (88-551-CM) supports the proliferation and activation of lymphocytes, including tumor infiltrating lymphocytes (TIL), Cytokine-Induced Killer (CIK) cells and Natural Killer (NK) cells. There is technical data to support the use in CAR-T as well as several other important immunotherapy applications.
Specifications
Specifications
| Form | Liquid |
| Quantity | 1000 mL |