missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lymphotoxin-alpha/TNF-beta Antibody (074), Alexa Fluor™ 488, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Lymphotoxin-alpha/TNF-beta Monoclonal specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for Sandwich ELISA Detection.
Spezifikation
Spezifikation
| Antigen | Lymphotoxin-alpha/TNF-beta |
| Anwendungen | ELISA |
| Klassifikation | Monoclonal |
| Klon | 74 |
| Konjugat | Alexa Fluor 488 |
| Verdünnung | Sandwich ELISA Detection |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | LT, LT-alpha, lymphotoxin alpha (TNF superfamily, member 1), TNFBlymphotoxin-alpha, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1 |
| Wirtsspezies | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Lymphotoxin-alpha/TNF-beta (P01374; Leu35-Leu205). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?