missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Lysobacter Enzymogenes Arg-C Recombinant Protein, PeproTech®

Artikelnummer. 17833923
Change view
Click to view available options
Menge:
12.5 x 20 μg
20 μg
25 x 20 μg
5 μg
5 x 20 μg
50 x 20 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
20 Mikrogramm
250 Mikrogramm
5 Mikrogramm
5 Stück
500 Mikrogramm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17833923 20 μg 20 Mikrogramm
17863973 5 x 20 μg 100 Mikrogramm
17814563 50 x 20 μg 1 Milligramm
17874053 12.5 x 20 μg 250 Mikrogramm
17864773 25 x 20 μg 500 Mikrogramm
17814223 5 μg 5 Mikrogramm
30087758 5 x 20 μg 5 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17833923 Lieferant Gibco™ Lieferanten-Nr. 4505420UG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

450-54-100UG will be provided as multiple 5 μg (450-54-5UG) and 20 μg (450-54-20UG) vials. Recombinant Lysobacter Enzymogenes Arg-C is a 26.8 kDa protease consisting of 252 amino acid residues including a C-terminal His-Tag. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Arg-C specifically cleaves at the carboxyl side of Arginine residues. Arg-C has a sulfhydryl requirement; it is activated by dithiothreitol, cysteine, or other sulfhydryl-containing reagents. The presence of calcium ions is essential. The enzyme is inhibited by oxidizing agents and sulfhydryl reactants, and by Co^2+, Cu2+, Cd^2+, and heavy metal ions.
TRUSTED_SUSTAINABILITY

Spezifikation

Zur Verwendung mit (Anwendung) Control, Bioactivity
Zusammensetzung Protein with no preservative
Molekulargewicht 26.8 kDa
Name Lysobacter Enzymogenes Arg-C
Reinigungsverfahren Purified
Menge 20 μg
Quelle Insect cells
Lagerungsbedingungen -20°C
Kennzeichnung RUO
Endotoxin-Konzentration <1 EU/μg
Gebräuchliche Bezeichnung Arg-C
Biologische Aktivität The reaction is measured as an increase in absorbance at 253 nm resulting from the hydrolysis of N-benzoyl-L-arginine ethyl ester (BAEE).
Produkttyp Protein
Konjugat Unconjugated
Rekombinant Recombinant
Proteinmarkierung His-tag
Sequenz GVGDIGSSDY CEKDIVCRVK PSAEFLSASK SVARMVFTPK TGYTGYCSGT LLNNSNSPKR QLFWSAAHCI STQKVANTLQ TYWLYDATGC DNDTLSDKAV TLTGGATLLH SHATRDTLLL ELKSAPPSGA YYAGWNSSAI ATKGTAIEGI HHPSGDLKKY SLGSVTALSS TIDGKKPLTK VAWTTGVTEG GSSGSGLFTI SSTSGYQLRG GLYGGTSYCS APSDPDYYSQ LDGVWSSIKT YFSPHHHHHH HH
Inhalt und Lagerung -20°C
Aktivität The reaction is measured as an increase in absorbance at 253 nm resulting from the hydrolysis of N-benzoyl-L-arginine ethyl ester (BAEE).
Expressionssystem Insect cells
Form Lyophilized
Reinheits- oder Qualitätsgrad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.