missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MAdCAM-1 Antibody, R&D Systems™
Rat Monoclonal Antibody
N.V.
Spezifikation
Antigen | MAdCAM-1 |
---|---|
Klon | 200530 |
Verdünnung | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Anwendungen | ELISA |
Klassifikation | Monoclonal |
Beschreibung
MAdCAM-1 Monoclonal specifically detects MAdCAM-1 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Spezifikation
MAdCAM-1 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
hMAdCAM-1, MACAM1, MAdCAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1 | |
MADCAM1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
200530 | |
ELISA | |
Unconjugated | |
Rat | |
Mouse | |
NP_038619 | |
8174 | |
Mouse myeloma cell line NS0-derived recombinant mouse MAdCAM-1 Gln22-Thr365 (predicted) Accession # NP_038619 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human and mouse MAdCAM-1 in ELISAs. |
For Research Use Only