Learn More
Clifton Magnetic hotplate stirrer Clifton CHS-2E analogue ceramic plate 100 to 1,500rpm, ambient to 450C, stirring volumes up to 15L
Marke:  Clifton CHS-2E
Magnetic hotplate stirrer Clifton CHS-2E analogue ceramic plate 100 to 1,500rpm, ambient to 450C, stirring volumes up to 15L
Weitere Details : Netto-Gewicht : 7.00000kg
Artikelnummer. 15119232