missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human RNF12 GST (N-Term) Protein

Product Code. 18604171 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18604171 10 μg 10µg
18633621 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18604171 Supplier R&D Systems™ Supplier No. H00051132Q0110ug
 Restricted Item
Verfügbar ab: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 51132
Molecular Weight (g/mol) MolecularWeight-theroretical: 34.87 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias DKFZp686N06224, E3 ubiquitin-protein ligase RLIM, E3 ubiquitin-protein ligase RNF12, EC 6.3.2, EC 6.3.2.-, FLJ25923, FLJ41093, LIM domain-interacting RING finger protein, MGC15161, NY-REN-43, Renal carcinoma antigen NY-REN-43, RING finger LIM domain-binding protein, RING finger protein 12FLJ42887, ring finger protein, LIM domain interacting, ring zinc finger LIM domain binding protein, ring zinc finger protein NY-REN-43antigen, R-LIM, RNF12FLJ45986
Gene Symbol RLIM
Research Category Zinc Finger
Product Line Novus
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein RNF12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.