missing translation for 'onlineSavingsMsg'
Learn More

Mandelamidoxim, 97 %, Thermo Scientific™

Catalog Number p-7039259
Click to view available options
Menge:
2 g
10 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
24440-15-1
C8H10N2O2
166.18
MFCD00194542
GXYVISXLFBQBKH-UHFFFAOYSA-N
mandelamidoxime, 1z-n',2-dihydroxy-2-phenylethanimidamide, n',2-dihydroxy-2-phenylacetimidamide, benzeneethanimidamide, n',alpha-dihydroxy, 2-amino-2-hydroxyimino-1-phenylethan-1-ol, benzeneethanimidamide, n,alpha-dihydroxy-9ci, e-n',2-dihydroxy-2-phenylethanimidamide
9656867
N',2-Dihydroxy-2-phenylethanimidamid
C1=CC=C(C=C1)C(C(=NO)N)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
24440-15-1
C8H10N2O2
166.18
MFCD00194542
GXYVISXLFBQBKH-UHFFFAOYSA-N
mandelamidoxime, 1z-n',2-dihydroxy-2-phenylethanimidamide, n',2-dihydroxy-2-phenylacetimidamide, benzeneethanimidamide, n',alpha-dihydroxy, 2-amino-2-hydroxyimino-1-phenylethan-1-ol, benzeneethanimidamide, n,alpha-dihydroxy-9ci, e-n',2-dihydroxy-2-phenylethanimidamide
9656867
N',2-Dihydroxy-2-phenylethanimidamid
C1=CC=C(C=C1)C(C(=NO)N)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Mandelamidoxime
Schmelzpunkt 148°C to 152°C
Menge 2 g
Formelmasse 166.18
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'