Learn More
Invitrogen™ MAP2K1 (MEK1) [S218D] [S222D] Recombinant Human Protein
MAP2K1 (MEK1) kinase is a dual-specific protein kinase that functions in mitogen-activated protein kinase (MAPK) cascades, controlling cell growth and differentiation.
Marke: Invitrogen™ PR3944C
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MAP2K1 is activated by a wide variety of growth factors and cytokines and also by membrane-dependent depolarization and calcium influx. This mutant has been activated by subsituting serines 218 and 222 with aspartic acid to mimic the phosphorylated state.
Spezifikation
NP_002746, S218D, S222D | |
50mM Tris, pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% NP-40, 1mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
MAP2K1 (MEK1) S218D S222D | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
49.2kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
STE7 Kinase Family | |
Recombinant | |
Baculovirus |