missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MAP2K4 (Human) Recombinant Protein
Human MAP2K4 full-length ORF ( AAH36032.1, 1 a.a. - 399 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00006416-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: MAAPSPSGGGGSGGGRGSGTPGPVGSPAPGHPAVSSMQGKRKALKLNFANPPFKSTARFTLNPNPTGVQNPHIERLRTHSIESSGKLKISPEQHWDFTAEDLKDLGEIGRGAYGSVNKMVHKPSGQIMAVKRIRSTVDEKEQKQLLMDLDVVMRSSDCPYIVQFYGALFREGDCWICMELMSTSFDKFYKYVYSVLDDVIPEEILGKITLATVKALNHLKENLKIIHRDIKPSNILLDRSGNIKLCDFGISGQLVDSIAKTRDAGCRPYMAPERIDPSASRQGYDVRSDVWSLGITLYELATGRFPYPKWNSVFDQLTQVVKGDPPQLSNSEEREFSPSFINFVNLCLTKDESKRPKYKELLKHPFILMYEERAVEVACYVCKILDQMPATPSSPMYVD
Spezifikation
AAH36032.1 | |
mitogen-activated protein kinase kinase 4 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAP2K4 | |
GST |
6416 | |
Wheat germ expression system | |
10 μg | |
JNKK, JNKK1, MAPKK4, MEK4, MKK4, PRKMK4, SEK1, SERK1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |