missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP2K6 (MKK6) Recombinant Human Protein
MAP2K6 (MKK6) is a serine/threonine kinase.
Marke: Invitrogen™ PR3950A
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MAP2K6 (MKK6) functions as an activator of p38 in the MAPK pathway.
Spezifikation
NP_002749 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
MAP2K6 (MKK6) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
41.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
STE7 Kinase Family | |
Recombinant | |
Baculovirus |