missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MAPK3 (ERK1) (Inactive), GST Tag Recombinant Protein
Human MAPK3 (ERK1) (Inactive), GST Tag Recombinant Protein
Marke: Invitrogen PV3312
Weitere Details : Netto-Gewicht : 3.36000kg
Beschreibung
MAPK3 (ERK1) is a serine⁄threonine kinase that is an important activator of p90, RSK, MSK, ELK1, and Stat3, and is involved in the MAPK pathway.Spezifikation
NP_002737 | |
5595 | |
100 μg | |
MAP Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
E. coli | |
Full Length, Recombinant, Inactive | |
Serine/Threonine Kinases |
Kinase Assay | |
70.8 kDa | |
MAPK3 (ERK1) | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: CMGC Group | |
mitogen-activated protein kinase 3 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.