missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10036-2-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
MEK5 and ERK5, a member of the MAP kinase subfamily of the Ser/Thr protein kinases, interact specifically with one another and not with MEK1/ERK1 or MEK2/ERK2 pathways. ERK5 is activated by tyrosine and threonine phosphorylation It is expressed in many adult tissues, abundantly in heart, placenta, lung, kidney and skeletal muscle, but is not detectable in liver. The second proline-rich region may interact with actin targeting the kinase to a specific location in the cell. ERK5 is autophosphorylated on threonine and tyrosine residues when the C-terminal part of the kinase, which could have a regulatory role, is absent.Spezifikation
MAPK7 | |
Polyclonal | |
Unconjugated | |
MAPK7 | |
Big MAP kinase 1, BMK 1, BMK1, ERK 5, ERK4, ERK5, MAP kinase 7, MAPK 7, MAPK7, PRKM7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23939, 5598 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13164, Q9WVS8 | |
MAPK7 | |
MAPK7 Fusion Protein Ag0017 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |