missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAPKAPK3 Recombinant Human Protein
MAPKAPK3 functions as an integrative element of signaling in both mitogen and stress responses.
Marke: Invitrogen™ PR4730B
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_004626 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
MAPKAPK3, 3PK, MAPKAP3 | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
47.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
MAPKAPK Kinase Family | |
Recombinant | |
Baculovirus |