missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Marapsin/Pancreasin Antibody, R&D Systems™
Goat Polyclonal Antibody
N.V.
Spezifikation
Antigen | Marapsin/Pancreasin |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 5 ug/mL |
Anwendungen | Western Blot, Immunoprecipitation, Neutralization |
Klassifikation | Polyclonal |
Beschreibung
Marapsin/Pancreasin Polyclonal specifically detects Marapsin/Pancreasin in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Spezifikation
Marapsin/Pancreasin | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 5 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CAP-2, CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPN, MPNMarapsin, Pancreasin, protease, serine 27, PRSS27, serine protease 27 | |
PRSS27 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation, Neutralization | |
Unconjugated | |
RUO | |
Q8BJR6 | |
83886 | |
Mouse myeloma cell line NS0-derived recombinant mouse Marapsin/Pancreasin Ala23-Thr290 Accession # Q8BJR6 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Marapsin/Pancreasin in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human Marapsin is observed. |
For Research Use Only