missing translation for 'onlineSavingsMsg'
Learn More

MCM7 Mouse anti-Human, Clone: MCM7/1469, Novus Biologicals™

Artikelnummer. 18213472 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μg
20 μg
Packungsgröße:
100 Mikrogramm
20 Mikrogramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18213472 100 μg 100 Mikrogramm
18218411 20 μg 20 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18213472 Lieferant Novus Biologicals Lieferanten-Nr. NBP259613100UG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

MCM7 Monoclonal specifically detects MCM7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen MCM7
Anwendungen Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page, Protein Array
Klassifikation Monoclonal
Klon MCM7/1469
Konzentration 0.2 mg/mL
Konjugat Unconjugated
Verdünnung Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin 0.5-1 ug/ml, SDS-Page, Protein Array 1:100-1:2000
Zusammensetzung 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gen-Alias CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47
Gensymbole MCM7
Wirtsspezies Mouse
Immunogen Recombinant human MCM7 protein fragment (aa195-319) (exact sequence is proprietary)
Molekulargewicht des Antigens 88 kDa
Reinigungsverfahren Protein A or G purified
Menge 100 μg
Regulatorischer Status RUO
Forschungsgebiet Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primär oder sekundär Primary
Gen-ID (Entrez) 4176
Testspezifität The specificity of this monoclonal antibody to its intended target was validated by HuProtTM Array, containing more than 19,000, full-length human proteins. MCM7 is one of the highly conserved mini-chromosome maintenance proteins (MCM) that is essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by theMCM proteins is a key component of the pre-replication complex and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 6 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. Cyclin D1-dependent kinase, CDK4, is found to associate with this protein, and may regulate the binding of this protein with the tumor suppressor protein RB1/RB.
Zielspezies Human
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Mehr anzeigen Weniger anzeigen

For Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.